Få mere at vide
(6Alpha)-21-(Acetyloxy)-17-hydroxy-6-methylpregna-1,4,9(11)-triene-3,20-dione, TRC
Artikelnummer. 30007420
Skift visning
Mængde:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30007420 50 mg
30011981 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30007420 Leverandør Toronto Research Chemicals Leverandørnr. TRCA16760050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 93239-37-3
Molekylær formel C24H30O5
Molekylvægt (g/mol) 398.49
Synonym 17,21-Dihydroxy-6alpha-methyl-pregna-1,4,9(11)-triene-3,20-dione 21-Acetate, NSC 86007
IUPAC navn [2-(17-hydroxy-6,10,13-trimethyl-3-oxo-7,8,12,14,15,16-hexahydro-6H-cyclopenta[a]phenanthren-17-yl)-2-oxoethyl] acetate
SMIL C[C@H]1C[C@H]2[C@@H]3CC[C@](O)(C(=O)COC(=O)C)[C@@]3(C)CC=C2[C@@]4(C)C=CC(=O)C=C14
Tekniske data
Kemisk navn eller materiale (6alpha)-21-(Acetyloxy)-17-hydroxy-6-methylpregna-1,4,9(11)-triene-3,20-dione
Mængde 50 mg
InChI formel InChI=1S/C24H30O5/c1-14-11-17-18(22(3)8-5-16(26)12-20(14)22)6-9-23(4)19(17)7-10-24(23,28)21(27)13-29-15(2)25/h5-6,8,12,14,17,19,28H,7,9-11,13H2,1-4H3
Formel vægt 398.2093