Få mere at vide
(6Alpha,11Beta,16Alpha)-21-(Acetyloxy)-6,9-difluoro-11,16,17-trihydroxypregna-1,4-diene-3,20-dione, TRC
Artikelnummer. 30327173
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30327173 50 mg
30320153 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30327173 Leverandør Toronto Research Chemicals Leverandørnr. TRCA36590350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (6alpha,11beta,16alpha)-21-(Acetyloxy)-6,9-difluoro-11,16,17-trihydroxypregna-1,4-diene-3,20-dione
CAS 4306-83-6
Molekylær formel C23H28F2O7
Mængde 50 mg
Synonym Fluocinolone Acetonide Impurity
InChI formel InChI=1S/C23H28F2O7/c1-11(26)32-10-19(30)23(31)17(28)8-13-14-7-16(24)15-6-12(27)4-5-20(15,2)22(14,25)18(29)9-21(13,23)3/h4-6,13-14,16-18,28-29,31H,7-10H2,1-3H3/t13-,14-,16-,17+,18-,20-,21-,22-,23-/m0/s1
SMIL CC(=O)OCC(=O)[C@@]1(O)[C@H](O)C[C@H]2[C@@H]3C[C@H](F)C4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C
IUPAC navn [2-[(6S,8S,9R,10S,11S,13S,14S,16R,17S)-6,9-difluoro-11,16,17-trihydroxy-10,13-dimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate
Molekylvægt (g/mol) 454.46
Formel vægt 454.1803