Få mere at vide
(6Alpha,11Beta,16Alpha,17Alpha)-6,9-Difluoro-17-[(2-furanylcarbonyl)oxy]-11-hydroxy-16-methyl-3-oxoandrosta-1,4-diene-17-carbothioic Acid (>85%), TRC
Artikelnummer. 30376302
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30376302 10 mg
30372743 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30376302 Leverandør Toronto Research Chemicals Leverandørnr. TRCD45025610MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 397864-40-3
Molekylær formel C26H28F2O6S
Molekylvægt (g/mol) 506.56
Synonym (6S,8S,9R,10S,11S,13S,14S,16R,17R)-6,9-Difluoro-17-((furan-2-carbonyl)oxy)-11-hydroxy-10,13,16-trimethyl-3-oxo-6,7,8,9,10,11,12,13,14,15,16,17-dodecahydro-3H-cyclopenta[a]phenanthrene-17-carbothioic S-Acid
IUPAC navn (6S,8S,9R,10S,11S,13S,14S,16R,17R)-6,9-difluoro-17-(furan-2-carbonyloxy)-11-hydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthrene-17-carbothioic S-acid
SMIL C[C@@H]1C[C@H]2[C@@H]3C[C@H](F)C4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(OC(=O)c5occc5)C(=O)S
Tekniske data
Kemisk navn eller materiale (6alpha,11beta,16alpha,17alpha)-6,9-Difluoro-17-[(2-furanylcarbonyl)oxy]-11-hydroxy-16-methyl-3-oxoandrosta-1,4-diene-17-carbothioic Acid
Mængde 10 mg
InChI formel InChI=1S/C26H28F2O6S/c1-13-9-15-16-11-18(27)17-10-14(29)6-7-23(17,2)25(16,28)20(30)12-24(15,3)26(13,22(32)35)34-21(31)19-5-4-8-33-19/h4-8,10,13,15-16,18,20,30H,9,11-12H2,1-3H3,(H,32,35)/t13-,15+,16+,18+,20+,23+,24+,25+,26+/m1/s1
Formel vægt 506.1575