Få mere at vide
Tocris Bioscience™ 6877002
Beskrivelse
6877002 is a CD40-TRAF6 interaction inhibitor. Suppresses CD40-induced IL-1β and IL-6 expression in macrophages in vitro. Improves insulin sensitivity, reduces adipose tissue inflammation, and ameliorates hepatosteatosis in diet-induced obese mice.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 3-[(2,5-Dimethylphenyl)amino]-1-phenyl-2-propen-1-one |
| CAS | 433249-94-6 |
| Mængde | 10 mg |
| Mål | Additional Cytokine Receptor Products |
| Molekylær formel | C17H17NO |
| Formel vægt | 251.32 |
| Renhed | >99% |