Få mere at vide
6-Thioguanosine Triphosphate Triethylamine Salt (>90%), TRC
Artikelnummer. 30452283
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30452283 10 mg
30452227 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30452283 Leverandør Toronto Research Chemicals Leverandørnr. TRCT35078510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 17670-19-8
Molekylær formel C10H16N5O13P3S.xC6H15N
Molekylvægt (g/mol) 539.25
Synonym 2-Amino-9-beta-D-ribofuranosyl-9H-purine-6(1H)-thione 5'-(Tetrahydrogen Triphosphate) Triethylamine Salt, 6-Mercapto-GTP, 6-Mercaptoguanosine 5'-Triphosphate Triethylamine Salt, 6-Thio-GTP, 6-Thioguanosine 5'-Triphosphate Triethylamine Salt, ((2R,3S,4R,5R)-5-(2-Amino-6-thioxo-1H-purin-9(6H)-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl Tetrahydrogen Triphosphate Triethylamine Salt
SMIL CCN(CC)CC.NC1=Nc2c(ncn2[C@@H]3O[C@H](COP(=O)(O)OP(=O)(O)OP(=O)(O)O)[C@@H](O)[C@H]3O)C(=S)N1
Tekniske data
Kemisk navn eller materiale 6-Thioguanosine Triphosphate Triethylamine Salt
Mængde 10 mg
InChI formel InChI=1S/C10H16N5O13P3S/c11-10-13-7-4(8(32)14-10)12-2-15(7)9-6(17)5(16)3(26-9)1-25-30(21,22)28-31(23,24)27-29(18,19)20/h2-3,5-6,9,16-17H,1H2,(H,21,22)(H,23,24)(H2,18,19,20)(H3,11,13,14,32)/t3-,5-,6-,9-/m1/s1