Få mere at vide
6-tert-Butyldimethylsilyloxy-4-(1-methylethenyl)-1-methyl-cyclohexane 1,2-Epoxide, TRC
Artikelnummer. 30344705
Skift visning
Mængde:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30344705 25 mg
30329492 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30344705 Leverandør Toronto Research Chemicals Leverandørnr. TRCB69136525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 6-tert-Butyldimethylsilyloxy-4-(1-methylethenyl)-1-methyl-cyclohexane 1,2-Epoxide
CAS 121289-19-8
Molekylær formel C16H30O2Si
Mængde 25 mg
Synonym (1,1-Dimethylethyl)dimethyl[[(1R,2S,4R,6R)-1-methyl-4-(1-methylethenyl)-7-oxabicyclo[4.1.0]hept-2-yl]oxy]-silane
InChI formel InChI=1S/C16H30O2Si/c1-11(2)12-9-13-16(6,17-13)14(10-12)18-19(7,8)15(3,4)5/h12-14H,1,9-10H2,2-8H3/t12-,13-,14+,16-/m1/s1
SMIL CC(=C)[C@H]1C[C@H](O[Si](C)(C)C(C)(C)C)[C@]2(C)O[C@@H]2C1
IUPAC navn tert-butyl-dimethyl-[[(1R,3R,5S,6R)-6-methyl-3-prop-1-en-2-yl-7-oxabicyclo[4.1.0]heptan-5-yl]oxy]silane
Molekylvægt (g/mol) 282.49
Formel vægt 282.2015