missing translation for 'onlineSavingsMsg'
Learn More

6-(tert-Butoxycarbonylamino)pyridine-3-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog Number p-4813240
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
910462-31-6
C16H25BN2O4
320.20
MFCD07781162
USZVCDRKIVKICD-UHFFFAOYSA-N
tert-butyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl carbamate, 2-tert-butoxycarbonylamino pyridine-5-boronic acid, pinacol ester, 2-boc-amino pyridine-5-boronic acid, pinacol ester, 2-boc-amino pyridine-5-boronic acid pinacol ester, tert-butyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-ylcarbamate, boc-6-aminopyridine-3-boronic acid pinacol ester, carbamic acid,n-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-pyridinyl-,1,1-dimethylethyl ester, tert-butyl n-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl carbamate, tert-butyl n-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl carbamate
17750222
tert-butyl N-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]carbamate
CC(C)(C)OC(=O)NC1=NC=C(C=C1)B1OC(C)(C)C(C)(C)O1
This item is not returnable. View return policy
910462-31-6
C16H25BN2O4
320.20
MFCD07781162
USZVCDRKIVKICD-UHFFFAOYSA-N
tert-butyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl carbamate, 2-tert-butoxycarbonylamino pyridine-5-boronic acid, pinacol ester, 2-boc-amino pyridine-5-boronic acid, pinacol ester, 2-boc-amino pyridine-5-boronic acid pinacol ester, tert-butyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-ylcarbamate, boc-6-aminopyridine-3-boronic acid pinacol ester, carbamic acid,n-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-pyridinyl-,1,1-dimethylethyl ester, tert-butyl n-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl carbamate, tert-butyl n-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl carbamate
17750222
tert-butyl N-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]carbamate
CC(C)(C)OC(=O)NC1=NC=C(C=C1)B1OC(C)(C)C(C)(C)O1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 203.0°C to 205.0°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 320.2
Percent Purity 97%
Chemical Name or Material 6-(tert-Butoxycarbonylamino)pyridine-3-boronic acid pinacol ester
compliance-icons
Product Identifier
  • 6-(tert-Butoxycarbonylamino)pyridine-3-boronic acid pinacol ester
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'