Få mere at vide
6-Sulfatoxy Melatonin Sodium Salt, TRC
Artikelnummer. 30008835
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30008835 10 mg
30008074 5 mg
30004731 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30008835 Leverandør Toronto Research Chemicals Leverandørnr. TRCS68905010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 76290-78-3
Molekylær formel C13H15N2NaO6S
Molekylvægt (g/mol) 350.32
Synonym N-[2-[5-Methoxy-6-(sulfooxy)-1H-indol-3-yl]ethyl]acetamide Sodium, 6-(Sulfonyloxy)melatonin Sodium, 6-Hydroxymelatonin Sulfate Sodium, 6-Sulphatoxymelatonin Sodium;
IUPAC navn sodium;[3-(2-acetamidoethyl)-5-methoxy-1H-indol-6-yl] sulfate
SMIL CC(=O)NCCC1=CNC2=CC(=C(C=C21)OC)OS(=O)(=O)[O-].[Na+]
Tekniske data
Kemisk navn eller materiale 6-Sulfatoxy Melatonin Sodium Salt
Renhedsgrad noter HPLC
Mængde 10 mg
InChI formel InChI=1S/C13H16N2O6S.Na/c1-8(16)14-4-3-9-7-15-11-6-13(21-22(17,18)19)12(20-2)5-10(9)11;/h5-7,15H,3-4H2,1-2H3,(H,14,16)(H,17,18,19);/q;+1/p-1
Formel vægt 350.055
Procent renhed >95%