Få mere at vide
6'-Sialyllactose Sodium Salt, TRC
Artikelnummer. 30409891
Skift visning
Mængde:
10 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30409891 250 mg
30412966 50 mg
30422172 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30409891 Leverandør Toronto Research Chemicals Leverandørnr. TRCS397405250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 157574-76-0
Molekylær formel C23H38NNaO19
Molekylvægt (g/mol) 655.53
Synonym 6'-Sialyllactose sodium, 6'-Sialyllactose Sodium Salt, D-Glucose, O-(N-acetyl-α-neuraminosyl)-(2→6)-O-β-D-galactopyranosyl-(1→4)-, monosodium salt (9CI), 6'-Sialyllactose sodium salt, D-Glucose, O-(N-acetyl-α-neuraminosyl)-(2→6)-O-β-D-galactopyranosyl-(1→4)-, sodium salt (1:1), O-(N-Acetyl-alpha-neuraminosyl)-(2-6)-O-beta-D-galactopyranosyl-(1-4)-D-glucose Sodium Salt
IUPAC navn sodium;(2R,4S,5R,6R)-5-acetamido-4-hydroxy-6-[(1R,2R)-1,2,3-trihydroxypropyl]-2-[[(2R,3R,4S,5R,6S)-3,4,5-trihydroxy-6-[(2R,3R,4R,5R)-1,2,4,5-tetrahydroxy-6-oxohexan-3-yl]oxyoxan-2-yl]methoxy]oxane-2-carboxylate
SMIL [Na+].CC(=O)N[C@@H]1[C@@H](O)C[C@@](OC[C@H]2O[C@@H](O[C@H]([C@H](O)CO)[C@H](O)[C@@H](O)C=O)[C@H](O)[C@@H](O)[C@H]2O)(O[C@H]1[C@H](O)[C@H](O)CO)C(=O)[O-]
Tekniske data
Kemisk navn eller materiale 6'-Sialyllactose Sodium Salt
Mængde 250 mg
InChI formel InChI=1S/C23H39NO19.Na/c1-7(28)24-13-8(29)2-23(22(38)39,43-20(13)15(34)10(31)4-26)40-6-12-16(35)17(36)18(37)21(41-12)42-19(11(32)5-27)14(33)9(30)3-25;/h3,8-21,26-27,29-37H,2,4-6H2,1H3,(H,24,28)(H,38,39);/q;+1/p-1/t8-,9-,10+,11+,12+,13+,14+,15+,16-,17-,18+,19+,20+,21-,23+;/m0./s1
Formel vægt 655.1936