Få mere at vide
[6]-Shogaol (>90%), TRC
Artikelnummer. 30012916
Skift visning
Mængde:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30012916 5 mg
30007679 10 mg
30008620 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30012916 Leverandør Toronto Research Chemicals Leverandørnr. TRCS3578005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 555-66-8
Molekylær formel C17 H24 O3
Molekylvægt (g/mol) 276.37
Synonym 1-(4-Hydroxy-3-methoxyphenyl)-4-decen-3-one, Shogaol;
IUPAC navn (E)-1-(4-hydroxy-3-methoxyphenyl)dec-4-en-3-one
SMIL CCCCC\C=C\C(=O)CCc1ccc(O)c(OC)c1
Tekniske data
Kemisk navn eller materiale [6]-Shogaol
Mængde 5 mg
InChI formel InChI=1S/C17H24O3/c1-3-4-5-6-7-8-15(18)11-9-14-10-12-16(19)17(13-14)20-2/h7-8,10,12-13,19H,3-6,9,11H2,1-2H3/b8-7+
Formel vægt 276.1725