missing translation for 'onlineSavingsMsg'
Få mere at vide

6-Propylamino-7-deazapurine, TRC

Artikelnummer. 30421121
Change view
Click to view available options
Mængde:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30421121 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30421121 Supplier Toronto Research Chemicals Supplier No. TRCP833250200MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 6-Propylamino-7-deazapurine
CAS 60972-21-6
Molekylær formel C9 H12 N4
Mængde 200 mg
InChI formel InChI=1S/C9H12N4/c1-2-4-10-8-7-3-5-11-9(7)13-6-12-8/h3,5-6H,2,4H2,1H3,(H2,10,11,12,13)
SMIL CCCNc1[nH]cnc2nccc12
IUPAC navn N-propyl-3H-pyrrolo[2,3-d]pyrimidin-4-amine
Molekylvægt (g/mol) 176.22
Formel vægt 176.1062
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.