Få mere at vide
6-Prenyl-rac-naringenin, TRC
Artikelnummer. 30428366
Skift visning
Mængde:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30428366 50 mg
30420706 10 mg
30424356 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30428366 Leverandør Toronto Research Chemicals Leverandørnr. TRCP71290550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 68682-01-9
Molekylær formel C20H20O5
Molekylvægt (g/mol) 340.37
Synonym 2,3-Dihydro-5,7-dihydroxy-2-(4-hydroxyphenyl)-6-(3-methyl-2-butenyl)-4H-1-benzopyran-4-one, (±)-2,3-Dihydro-5,7-dihydroxy-2-(4-hydroxyphenyl)-6-(3-methyl-2-buten-1-yl)-4H-1-benzopyran-4-one, (±)-6-Prenylnaringenin, 4H-1-Benzopyran-4-one, 2,3-dihydro-5,7-dihydroxy-2-(4-hydroxyphenyl)-6-(3-methyl-2-buten-1-yl)-, 4H-1-Benzopyran-4-one, 2,3-dihydro-5,7-dihydroxy-2-(4-hydroxyphenyl)-6-(3-methyl-2-butenyl)- (9CI), 2,3-Dihydro-5,7-dihydroxy-2-(4-hydroxyphenyl)-6-(3-methyl-2-buten-1-yl)-4H-1-benzopyran-4-one
IUPAC navn 5,7-dihydroxy-2-(4-hydroxyphenyl)-6-(3-methylbut-2-enyl)-2,3-dihydrochromen-4-one
SMIL CC(=CCc1c(O)cc2OC(CC(=O)c2c1O)c3ccc(O)cc3)C
Tekniske data
Kemisk navn eller materiale 6-Prenyl-rac-naringenin
Mængde 50 mg
InChI formel InChI=1S/C20H20O5/c1-11(2)3-8-14-15(22)9-18-19(20(14)24)16(23)10-17(25-18)12-4-6-13(21)7-5-12/h3-7,9,17,21-22,24H,8,10H2,1-2H3
Formel vægt 340.1311