Learn More
SCHOTT AG Lighting and Imaging™ 6-Point Ring Light
Provides optimal lighting for contrast while reducing shadows. Mounts directly to objective by means of thumb screw. For use with Cold light source KL300LED
Brand: SCHOTT AG Lighting and Imaging™ 155102
Additional Details : Weight : 5.00000kg
Description
- 6-point Ringlight with eco-friendly Puravis fiber optics
- Fiber bundle protected with flexible metal PVC sheathing.
- Housing made of rugged, black anodized aluminum.
- Cost effective in comparison to annular ringlights
- Illumination with reduced shadows, but still sufficient shadows for improved contrasts
Specifications
Fiber Optic Ringlight | |
Stereo Microscopy illumination | |
23 mm | |
Eco-friendly Puravis Glass Optical Fibers |
66 mm ID | |
Cold light source KL300LED | |
1000 mm |