Få mere at vide
6-Perdeoxy-6-perchloro-Gamma-cyclodextrin, >90%, TRC
Artikelnummer. 30416474
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30416474 100 mg
30411194 50 mg
30413019 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30416474 Leverandør Toronto Research Chemicals Leverandørnr. TRCP999728100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 173094-60-5
Molekylær formel C48H72Cl8O32
Molekylvægt (g/mol) 1444.69
Synonym 6A,6B,6C,6D,6E,6F,6G,6H-Octachloro-6A,6B,6C,6D,6E,6F,6G,6H-octadeoxy-gamma-cyclodextrin
SMIL O[C@@H]1[C@@H](O)[C@@H]2O[C@H]3O[C@H](CCl)[C@@H](O[C@H]4O[C@H](CCl)[C@@H](O[C@H]5O[C@H](CCl)[C@@H](O[C@H]6O[C@H](CCl)[C@@H](O[C@H]7O[C@H](CCl)[C@@H](O[C@H]8O[C@H](CCl)[C@@H](O[C@H]9O[C@H](CCl)[C@@H](O[C@H]1O[C@@H]2CCl)[C@H](O)[C@H]9O)[C@H](O)[C@H]8O)[C@H](O)[C@H]7O)[C@H](O)[C@H]6O)[C@H](O)[C@H]5O)[C@H](O)[C@H]4O)[C@H](O)[C@H]3O
Tekniske data
Kemisk navn eller materiale 6-Perdeoxy-6-perchloro-gamma-cyclodextrin
Mængde 100 mg
InChI formel InChI=1S/C48H72Cl8O32/c49-1-9-33-17(57)25(65)41(73-9)82-34-10(2-50)75-43(27(67)19(34)59)84-36-12(4-52)77-45(29(69)21(36)61)86-38-14(6-54)79-47(31(71)23(38)63)88-40-16(8-56)80-48(32(72)24(40)64)87-39-15(7-55)78-46(30(70)22(39)62)85-37-13(5-53)76-44(28(68)20(37)60)83-35-11(3-51)74-42(81-33)26(66)18(35)58/h9-48,57-72H,1-8H2/t9-,10-,11-,12-,13-,14-,15-,16-,17-,18-,19-,20-,21-,22-,23-,24-,25-,26-,27-,28-,29-,30-,31-,32-,33-,34-,35-,36-,37-,38-,39-,40-,41-,42-,43-,44-,45-,46-,47-,48-/m1/s1
Formel vægt 1440.1515