Få mere at vide
6-Oxo DL-Norleucine, Formate Salt, TRC
Artikelnummer. 30011315
Skift visning
Mængde:
1 mg
2.5 mg
5 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30011315 5 mg -
30004421 2.5 mg 2.5mg
30004965 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30011315 Leverandør Toronto Research Chemicals Leverandørnr. TRCO8701405MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2125941-25-3
Molekylær formel C6H11NO3 •x(HCO2H)
Molekylvægt (g/mol) 145.164603
Synonym 6-Oxo-norleucine, 5-Formylnorvaline, 2-Amino-5-formylvaleric Acid, Allysine, α-Aminoadipic Acid δ-Semialdehyde, α-Aminoadipic δ-Semialdehyde;
SMIL O=CCCCC(N)C(O)=O>>OC(C1=NCCCC1)=O.O
Tekniske data
Kemisk navn eller materiale 6-Oxo DL-Norleucine, Formate Salt
InChI formel InChI=1S/C6H11NO3/c7-5(6(9)10)3-1-2-4-8/h4-5H,1-3,7H2,(H,9,10)