Learn More
Alfa Aesar™ 6-O-tert-Butyldiphenylsilyl-D-glucal, 97%
Brand: Alfa Aesar™ H60191.MD
Additional Details : CAS Number : 87316-22-1 Weight : 0.00025kg
Quantity | 250mg |
---|
Chemical Identifiers
87316-22-1 | |
C22H28O4Si | |
MFCD01321278 | |
6-o-tert-butyldiphenylsilyl-d-glucal, 6-o-tert-butyldiphenylsilyl-d-glucose, 6-o-tert-butyl diphenyl silyl-d-glucose, 2r,3s,4r,5r-6-tert-butyldiphenylsilyl oxy-2,3,4,5-tetrahydroxyhexanal |
Specifications
6-O-tert-Butyldiphenylsilyl-D-glucal | |
-9°C | |
87316-22-1 | |
MFCD01321278 | |
384.54 |
+9° (c=1.1 in Chloroform) | |
250mg | |
C22H28O4Si | |
6-o-tert-butyldiphenylsilyl-d-glucal, 6-o-tert-butyldiphenylsilyl-d-glucose, 6-o-tert-butyl diphenyl silyl-d-glucose, 2r,3s,4r,5r-6-tert-butyldiphenylsilyl oxy-2,3,4,5-tetrahydroxyhexanal | |
97% |