Få mere at vide
6-O-Desmethyl Moxonidine, TRC
Artikelnummer. 30135953
Skift visning
Mængde:
2.5 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30135953 2.5 mg
30004196 10 mg
30011529 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30135953 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2919402.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 352457-33-1
Molekylær formel C8 H10 Cl N5 O
Molekylvægt (g/mol) 227.65
Synonym 4(1H)-Pyrimidinone, 6-chloro-5-[(4,5-dihydro-1H-imidazol-2-yl)amino]-2-methyl-, 6-Chloro-5-[(4,5-dihydro-1H-imidazol-2-yl)amino]-2-methyl-4(1H)-pyrimidinone, 6-Chloro-5-[(imidazolidin-2-ylidene)-amino]-2-methylpyrimidin-4-ol, 6-Desmethylmoxonidine
IUPAC navn 6-chloro-5-(imidazolidin-2-ylideneamino)-2-methylpyrimidin-4-ol
SMIL Cc1nc(O)c(N=C2NCCN2)c(Cl)n1
Tekniske data
Kemisk navn eller materiale 6-O-Desmethyl Moxonidine
Mængde 2.5 mg
InChI formel InChI=1S/C8H10ClN5O/c1-4-12-6(9)5(7(15)13-4)14-8-10-2-3-11-8/h2-3H2,1H3,(H2,10,11,14)(H,12,13,15)
Formel vægt 227.0574