Få mere at vide
6-O-Desmethyl Donepezil, TRC
Artikelnummer. 30004282
Skift visning
Mængde:
1 mg
2.5 mg
5 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30004282 1 mg -
30005015 5 mg -
30007641 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30004282 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2920151MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 120013-56-1
Molekylær formel C23H27NO3
Molekylvægt (g/mol) 365.47
Synonym 1H-Inden-1-one, 2,3-dihydro-6-hydroxy-5-methoxy-2-[[1-(phenylmethyl)-4-piperidinyl]methyl]-, 2,3-Dihydro-6-hydroxy-5-methoxy-2-[[1-(phenylmethyl)-4-piperidinyl]methyl]-1H-inden-1-one, 1-Benzyl-4-[(6-hydroxy-5-methoxy-1-oxo-indan-2-yl)methyl]piperidine, 6-O-Desmethyldonepezil
SMIL COc1cc2CC(CC3CCN(Cc4ccccc4)CC3)C(=O)c2cc1O
Tekniske data
Kemisk navn eller materiale 6-O-Desmethyl Donepezil
Mængde 1 mg
InChI formel InChI=1S/C23H27NO3/c1-27-22-13-18-12-19(23(26)20(18)14-21(22)25)11-16-7-9-24(10-8-16)15-17-5-3-2-4-6-17/h2-6,13-14,16,19,25H,7-12,15H2,1H3
Formel vægt 365.1991