Få mere at vide
6-Nitrodopamine, TRC
Artikelnummer. 30132727
Skift visning
Mængde:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30132727 250 mg
30011902 100 mg
30007206 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30132727 Leverandør Toronto Research Chemicals Leverandørnr. TRCN493720250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 21581-49-7
Molekylær formel C8H10N2O4
Molekylvægt (g/mol) 198.18
Synonym 4-(2-Aminoethyl)-5-nitropyrocatechol, 4-(2-Aminoethyl)-5-nitro-1,2-benzenediol, 4-(2-Aminoethyl)-5-nitrobenzene-1,2-diol
SMIL CC1c2cc3C(C)(C)C(C)C(C)(C)c3cc2COC1
Tekniske data
Kemisk navn eller materiale 6-Nitrodopamine
Mængde 250 mg
InChI formel InChI=1S/C8H10N2O4/c9-2-1-5-3-7(11)8(12)4-6(5)10(13)14/h3-4,11-12H,1-2,9H2