Få mere at vide
6-Nitrochrysene-d11, CDN
Artikelnummer. 30321757
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30321757 0.01 g
30319851 0.005 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321757 Leverandør CDN Leverandørnr. CDND43600.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 6-Nitrochrysene (D11)
CAS 203805-92-9
Molekylær formel C18 D11 N O2
Mængde 0.01 g
InChI formel InChI=1 S/C18H11NO2/c20-19(21)18-11-17-13-6-2-1-5-12(13)9-10-15(17)14-7-3-4-8-16(14)18/h1-11 H/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D,9 D,10 D,11 D
SMIL [2 H]c1c([2 H])c([2 H])c2c(c1[2 H])c([2 H])c([2 H])c3c4c([2 H])c([2 H])c([2 H])c([2 H])c4c(c([2 H])c23)[N+](=O)[O-]
IUPAC navn 1,2,3,4,5,6,7,8,9,10,11-undecadeuterio-12-nitro-chrysene
Molekylvægt (g/mol) 284.35
Formel vægt 284.148 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity