missing translation for 'onlineSavingsMsg'
Learn More

6-Nitro-2H-1,4-benzothiazin-3(4H)-one, 97%, Thermo Scientific™

Catalog Number p-7058139
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
21762-74-3
C8H6N2O3S
210.21
MFCD00456556
BALSKAVZBPPKCS-UHFFFAOYSA-N
6-nitro-2h-benzo b 1,4 thiazin-3 4h-one, 6-nitro-2h-1,4-benzothiazin-3 4h-one, 2h-1,4-benzothiazin-3 4h-one, 6-nitro, 6-nitro-2,4-dihydro-1,4-benzothiazin-3-one, 2h-1,4-benzothiazin-3 4h-one,6-nitro, 6-nitro-2h-benzo b-1,4 thiazin-3 4h-one, 6-nitro-3,4-dihydro-2h-1,4-benzothiazin-3-one
4182356
6-nitro-3,4-dihydro-2H-1,4-benzothiazin-3-one
[O-][N+](=O)C1=CC2=C(SCC(=O)N2)C=C1
This item is not returnable. View return policy
21762-74-3
C8H6N2O3S
210.21
MFCD00456556
BALSKAVZBPPKCS-UHFFFAOYSA-N
6-nitro-2h-benzo b 1,4 thiazin-3 4h-one, 6-nitro-2h-1,4-benzothiazin-3 4h-one, 2h-1,4-benzothiazin-3 4h-one, 6-nitro, 6-nitro-2,4-dihydro-1,4-benzothiazin-3-one, 2h-1,4-benzothiazin-3 4h-one,6-nitro, 6-nitro-2h-benzo b-1,4 thiazin-3 4h-one, 6-nitro-3,4-dihydro-2h-1,4-benzothiazin-3-one
4182356
6-nitro-3,4-dihydro-2H-1,4-benzothiazin-3-one
[O-][N+](=O)C1=CC2=C(SCC(=O)N2)C=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 210.21
Percent Purity 97%
Chemical Name or Material 6-Nitro-2H-1,4-benzothiazin-3(4H)-one
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'