Få mere at vide
6-Methylthioguanine-d3, TRC
Artikelnummer. 30473244
Skift visning
Mængde:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30473244 250 mg
30469541 500 mg
30468073 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30473244 Leverandør Toronto Research Chemicals Leverandørnr. TRCM330722250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1346601-96-4
Molekylær formel C6 2H3 H4 N5 S
Molekylvægt (g/mol) 184.24
Synonym 6-(Trideuteriomethylsulfanyl)-7H-purin-2-amine, 6-Methylthioguanine-d3, 6-Methylthioguanine-D3 (methyl-D3), 2-Amino-6-(methyl-d3)thiopurine, 6-Trideuteromethylthioguanine
IUPAC navn 6-(trideuteriomethylsulfanyl)-7H-purin-2-amine
SMIL [2H]C([2H])([2H])Sc1nc(N)nc2nc[nH]c12
Tekniske data
Kemisk navn eller materiale 6-Methylthioguanine methyl-d3
Mængde 250 mg
InChI formel InChI=1S/C6H7N5S/c1-12-5-3-4(9-2-8-3)10-6(7)11-5/h2H,1H3,(H3,7,8,9,10,11)/i1D3
Formel vægt 184.061