Få mere at vide
6-Methylquinoline, TRC
Artikelnummer. 30479728
Skift visning
Mængde:
10 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30479728 10 g
30464770 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30479728 Leverandør Toronto Research Chemicals Leverandørnr. TRCM32800510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 91-62-3
Molekylær formel C10 H9 N
Molekylvægt (g/mol) 143.185
Synonym 6-Methylquinoline, NSC 4152
IUPAC navn 6-methylquinoline
SMIL Cc1ccc2ncccc2c1
Tekniske data
Kemisk navn eller materiale 6-Methylquinoline
Mængde 10 g
InChI formel InChI=1S/C10H9N/c1-8-4-5-10-9(7-8)3-2-6-11-10/h2-7H,1H3
Formel vægt 143.0735