missing translation for 'onlineSavingsMsg'
Learn More

6-Methylnicotinamide, 98%, Thermo Scientific Chemicals

Catalog Number p-7023019
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
6960-22-1
C7H8N2O
136.154
MFCD00006342
IJXDURUAYOKSIS-UHFFFAOYSA-N
96351
6-methylpyridine-3-carboxamide
CC1=NC=C(C=C1)C(=O)N
This item is not returnable. View return policy
6960-22-1
C7H8N2O
136.154
MFCD00006342
IJXDURUAYOKSIS-UHFFFAOYSA-N
96351
6-methylpyridine-3-carboxamide
CC1=NC=C(C=C1)C(=O)N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 195°C to 199°C
Quantity 1 g
Beilstein 112050
Synonym 6-methylnicotinamide, 6-methyl nicotinamide, 3-pyridinecarboxamide, 6-methyl, 6-methyl-3-pyridinecarboxamide, 8ej, nicotinamide, 6-methyl, 3-carbamoyl-6-methylpyridine, ksc630k7f, 2-methylpyridine-5-carboxamide, 3-pyridinecarboxamide,6-methyl
Formula Weight 136.15
Percent Purity 98%
Chemical Name or Material 6-Methylnicotinamide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'