Få mere at vide
6-Methylmercaptopurine-d3, TRC
Artikelnummer. 30470096
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30470096 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30470096 Leverandør Toronto Research Chemicals Leverandørnr. TRCM32027225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 6-Methylmercaptopurine-D3
CAS 33312-93-5
Renhedsgrad noter HPLC
Molekylær formel C6 D3 H3 N4 S
Mængde 25 mg
Synonym 9H-Purine','6-(methyl-d3-thio)- (ACI), 1H-Purine','6-(methyl-d3-thio)- (8CI','9CI), 6-(Methyl-d3-thio)-9H-purine (ACI), 6-(Methylthio-d3)purine, 6-Methyl-d3-mercaptopurine, 6-Methylmercaptopurine-d3, 6-Methylmercaptopurine-D3 (S-methyl-D3), S-(Methyl-d3)-6-mercaptopurine
InChI formel InChI=1S/C6H6N4S/c1-11-6-4-5(8-2-7-4)9-3-10-6/h2-3H,1H3,(H,7,8,9,10)/i1D3
SMIL [2H]C([2H])([2H])Sc1ncnc2nc[nH]c12
IUPAC navn 6-(trideuteriomethylsulfanyl)-7H-purine
Molekylvægt (g/mol) 169.2221
Formel vægt 169.0501
Procent renhed >95