Learn More
6-Methylchroman-4-one, 95%, BLD Pharm™
Product Code. 30979685
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
10 g
25 g
Unit Size:
25g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30979685 5 g -
30974539 10 g -
30975635 250 mg -
30978068 25 g 25g
30975252 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 30979685 Supplier BLD Pharm Supplier No. BD118295g
 Restricted Item
Forventet forsendelse: 27-11-2026
Specifications
CAS 39513-75-2
Molecular Formula C10H10O2
MDL Number MFCD00024071
Quantity 5 g
SMILES O=C1CCOC2=C1C=C(C)C=C2
Molecular Weight (g/mol) 162.19
Percent Purity 95%
Chemical Name or Material 6-Methylchroman-4-one