Få mere at vide
6-Methyl-N-[2-(4-sulphamoylphenyl)ethyl]pyrazine-2-carboxamide, Mikromol™
Artikelnummer. 30052396
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30052396 25 mg
17941996 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30052396 Leverandør Mikromol Leverandørnr. MM0942.040025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
6-Methyl-N-[2-(4-sulphamoylphenyl)ethyl]pyrazine-2-carboxamide
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 6-Methyl-N-[2-(4-sulphamoylphenyl)ethyl]pyrazine-2-carboxamide
CAS 1346600-54-1
Analyt- eller komponentnavne 6-Methyl-N-[2-(4-sulphamoylphenyl)ethyl]pyrazine-2-carboxamide
Forsendelsestilstand Room Temperature
Molekylær formel C14 H16 N4 O3 S
Certificeringer/compliance ISO 9001
Synonym N-[2-[4-(Aminosulfonyl)phenyl]ethyl]-6-methyl-2-pyrazinecarboxamide
InChI formel InChI=1S/C14H16N4O3S/c1-10-8-16-9-13(18-10)14(19)17-7-6-11-2-4-12(5-3-11)22(15,20)21/h2-5,8-9H,6-7H2,1H3,(H,17,19)(H2,15,20,21)
SMIL O=C(NCCC1=CC=C(C=C1)S(=O)(=O)N)C=2N=C(C=NC2)C
IUPAC navn 6-methyl-N-[2-(4-sulfamoylphenyl)ethyl]pyrazine-2-carboxamide
Molekylvægt (g/mol) 320.37
Mængde 25 mg
Formel vægt 320.0943 g/mol