Få mere at vide
6-Methyl-5-hepten-2-one-1,1,1,3,3-d5, TRC
Artikelnummer. 30479957
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30479957 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30479957 Leverandør Toronto Research Chemicals Leverandørnr. TRCM311902100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 6-Methyl-5-hepten-2-one-1,1,1,3,3-d5
CAS 55320-91-7
Molekylær formel C8 D5 H9 O
Mængde 100 mg
InChI formel InChI=1S/C8H14O/c1-7(2)5-4-6-8(3)9/h5H,4,6H2,1-3H3/i3D3,6D2
SMIL [2H]C([2H])([2H])C(=O)C([2H])([2H])CC=C(C)C
IUPAC navn 1,1,1,3,3-pentadeuterio-6-methylhept-5-en-2-one
Molekylvægt (g/mol) 131.227
Formel vægt 131.136