Få mere at vide
6-Methyl-4-phenylchroman-2-one, Mikromol™
Artikelnummer. 30051226
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30051226 100 mg
30050386 25 mg
30457766 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30051226 Leverandør Mikromol Leverandørnr. MM0900.04
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
6-Methyl-4-phenylchroman-2-one
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 6-Methyl-4-phenylchroman-2-one
CAS 40546-94-9
Analyt- eller komponentnavne 6-Methyl-4-phenylchroman-2-one
Forsendelsestilstand Room Temperature
Molekylær formel C16 H14 O2
Certificeringer/compliance ISO 17025
Synonym 2H-1-Benzopyran-2-one, 3,4-dihydro-6-methyl-4-phenyl-, Hydrocoumarin, 6-methyl-4-phenyl- (6CI,7CI), 3,4-Dihydro-6-methyl-4-phenyl-2H-benzopyran-2-one, 6-Methyl-4-phenyl-3,4-dihydro-1-benzopyran-2-one, 6-Methyl-4-phenyl-3,4-dihydrocoumarin, 6-Methyl-4-phenylchroman-2-one
InChI formel InChI=1S/C16H14O2/c1-11-7-8-15-14(9-11)13(10-16(17)18-15)12-5-3-2-4-6-12/h2-9,13H,10H2,1H3
SMIL Cc1ccc2OC(=O)CC(c3ccccc3)c2c1
IUPAC navn 6-methyl-4-phenyl-3,4-dihydrochromen-2-one
Molekylvægt (g/mol) 238.28
Mængde 100 mg
Formel vægt 238.0994 g/mol