Få mere at vide
6-Methyl-2-heptanone, TRC
Artikelnummer. 30134105
Skift visning
Mængde:
2.5 g
10 g
Pakningsstørrelse:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30134105 10 g -
30009073 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30134105 Leverandør Toronto Research Chemicals Leverandørnr. TRCM31191510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 928-68-7
Molekylær formel C8H16O
Molekylvægt (g/mol) 128.21
Synonym 2-Isooctanone, 2-Methyl-6-heptanone, 6-Methyl-2-heptanone, Isooctan-2-one, Methyl isohexyl ketone, NSC 39665
IUPAC navn 6-methylheptan-2-one
SMIL CC(C)CCCC(=O)C
Tekniske data
Kemisk navn eller materiale 6-Methyl-2-heptanone
Mængde 10 g
InChI formel InChI=1S/C8H16O/c1-7(2)5-4-6-8(3)9/h7H,4-6H2,1-3H3
Formel vægt 128.1201