Få mere at vide
6-Methyl-2-(4-methylphenyl)-imidazo[1,2-a]pyridine, TRC
Artikelnummer. 30465208
Skift visning
Mængde:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30465208 10 g
30480547 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30465208 Leverandør Toronto Research Chemicals Leverandørnr. TRCM32205010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 88965-00-8
Molekylær formel C15 H14 N2
Molekylvægt (g/mol) 222.29
Synonym Imidazo[1,2-a]pyridine, 6-methyl-2-(4-methylphenyl)-, 2-(4-Methylphenyl)-6-methylimidazo[1,2-a]pyridine
IUPAC navn 6-methyl-2-(4-methylphenyl)imidazo[1,2-a]pyridine
SMIL Cc1ccc(cc1)c2cn3cc(C)ccc3n2
Tekniske data
Kemisk navn eller materiale 6-Methyl-2-(4-methylphenyl)imidazo[1,2-a]pyridine
Mængde 10 g
InChI formel InChI=1S/C15H14N2/c1-11-3-6-13(7-4-11)14-10-17-9-12(2)5-8-15(17)16-14/h3-10H,1-2H3
Formel vægt 222.1157