Få mere at vide
6-Methyl-2-(4-methylphenyl)imidazo[1,2-a]pyridine, Mikromol™
Artikelnummer. 30050860
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30050860 25 mg
30051218 100 mg
30463060 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30050860 Leverandør Mikromol Leverandørnr. MM0333.150025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
6-Methyl-2-(4-methylphenyl)imidazo[1,2-a]pyridine
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 6-Methyl-2-(4-methylphenyl)imidazo[1,2-a]pyridine
CAS 88965-00-8
Analyt- eller komponentnavne 6-Methyl-2-(4-methylphenyl)imidazo[1,2-a]pyridine
Forsendelsestilstand Room Temperature
Molekylær formel C15 H14 N2
Certificeringer/compliance ISO 9001
Synonym Imidazo[1,2-a]pyridine, 6-methyl-2-(4-methylphenyl)-, 2-(4-Methylphenyl)-6-methylimidazo[1,2-a]pyridine
InChI formel InChI=1S/C15H14N2/c1-11-3-6-13(7-4-11)14-10-17-9-12(2)5-8-15(17)16-14/h3-10H,1-2H3
SMIL Cc1ccc(cc1)c2cn3cc(C)ccc3n2
IUPAC navn 6-methyl-2-(4-methylphenyl)imidazo[1,2-a]pyridine
Molekylvægt (g/mol) 222.29
Mængde 25 mg
Formel vægt 222.1157 g/mol