Få mere at vide
6-Methoxy-2-naphthol, TRC
Artikelnummer. 30476367
Skift visning
Mængde:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30476367 5 g
30466802 10 g
30472107 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30476367 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2628905G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 5111-66-0
Molekylær formel C11 H10 O2
Molekylvægt (g/mol) 174.2
Synonym 6-Methoxynaphthalen-2-ol, Naproxen Imp. H (EP), Hydroxynerolin
IUPAC navn 6-methoxynaphthalen-2-ol
SMIL COc1ccc2cc(O)ccc2c1
Tekniske data
Kemisk navn eller materiale 6-Methoxynaphthalen-2-ol (Hydroxynerolin)
Mængde 5 g
InChI formel InChI=1S/C11H10O2/c1-13-11-5-3-8-6-10(12)4-2-9(8)7-11/h2-7,12H,1H3
Formel vægt 174.0681