Få mere at vide
6-Methoxy-2-(4-methoxyphenyl)benzo[b]thiophene, Mikromol™
Artikelnummer. 30050241
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30050241 100 mg
30050205 25 mg
30460575 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30050241 Leverandør Mikromol Leverandørnr. MM0738.07
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
6-Methoxy-2-(4-methoxyphenyl)benzo[b]thiophene
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 6-Methoxy-2-(4-methoxyphenyl)benzo[b]thiophene
CAS 63675-74-1
Analyt- eller komponentnavne 6-Methoxy-2-(4-methoxyphenyl)benzo[b]thiophene
Forsendelsestilstand Room Temperature
Molekylær formel C16 H14 O2 S
Certificeringer/compliance ISO 17025
Synonym Benzo[b]thiophene, 6-methoxy-2-(4-methoxyphenyl)-, 2-(4-Methoxyphenyl)-6-methoxybenzo[b]thiophene, 6-Methoxy-2-(4-methoxyphenyl)benzo[b]thiophene
InChI formel InChI=1S/C16H14O2S/c1-17-13-6-3-11(4-7-13)15-9-12-5-8-14(18-2)10-16(12)19-15/h3-10H,1-2H3
SMIL COc1ccc(cc1)c2cc3ccc(OC)cc3s2
IUPAC navn 6-methoxy-2-(4-methoxyphenyl)-1-benzothiophene
Molekylvægt (g/mol) 270.35
Mængde 100 mg
Formel vægt 270.0715 g/mol