Learn More
6-Hydroxyquinoline, 98%, Thermo Scientific Chemicals
Catalog Number p-2690907
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
580-16-5
C9H7NO
145.16
MFCD00047611
OVYWMEWYEJLIER-UHFFFAOYSA-N
6-hydroxyquinoline, 6-quinolinol, 1h-1,6-epoxyquinoline, ccris 4331, 6-chinolinol, 6-hydroxyquinoline 6-quinolinol, quinoline-6-ol, 6-quinolinoloxine, 6-hydroxy-chinolin, 6-hydroxy-quinoline
11374
CHEBI:48994
quinolin-6-ol
C1=CC2=C(C=CC(=C2)O)N=C1
580-16-5
C9H7NO
145.16
MFCD00047611
OVYWMEWYEJLIER-UHFFFAOYSA-N
6-hydroxyquinoline, 6-quinolinol, 1h-1,6-epoxyquinoline, ccris 4331, 6-chinolinol, 6-hydroxyquinoline 6-quinolinol, quinoline-6-ol, 6-quinolinoloxine, 6-hydroxy-chinolin, 6-hydroxy-quinoline
11374
CHEBI:48994
quinolin-6-ol
C1=CC2=C(C=CC(=C2)O)N=C1
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 190°C to 195°C
Color Beige to Gray or Brown
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Beilstein 21, V,3, 244
Solubility Information Solubility in water: almost insoluble in water
Formula Weight 145.16
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 6-Hydroxyquinoline, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 6-Hydroxyquinoline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.