Få mere at vide
6-Hydroxy Melatonin-d4 (Major), TRC
Artikelnummer. 30408127
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30408127 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30408127 Leverandør Toronto Research Chemicals Leverandørnr. TRCH9446271MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 6-Hydroxy Melatonin-d4
CAS 69533-61-5
Renhedsgrad noter HPLC
Molekylær formel C13H12D4N2O3
Mængde 1 mg
Synonym N-[2-(6-Hydroxy-5-methoxy-1H-indol-3-yl)ethyl]acetamide-d4, 6-Hydroxy-N-acetyl-5-methoxytryptamine-d4
InChI formel InChI=1S/C13H16N2O3/c1-8(16)14-4-3-9-7-15-11-6-12(17)13(18-2)5-10(9)11/h5-7,15,17H,3-4H2,1-2H3,(H,14,16)/i3D2,4D2
SMIL CC(NC([2H])([2H])C([2H])([2H])C1=CNC2=C1C=C(OC)C(O)=C2)=O
Molekylvægt (g/mol) 252.3
Formel vægt 252.1412
Procent renhed >95