Få mere at vide
6-Hydroxy-L-DOPA, TRC
Artikelnummer. 30137106
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137106 250 mg
30008371 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30137106 Leverandør Toronto Research Chemicals Leverandørnr. TRCH941725250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 27244-64-0
Molekylær formel C9 H11 N O5
Molekylvægt (g/mol) 213.19
Synonym 3-(3,4,6-Trihydroxyphenyl)-alanine
IUPAC navn (2S)-2-amino-3-(2,4,5-trihydroxyphenyl)propanoic acid
SMIL N[C@@H](Cc1cc(O)c(O)cc1O)C(=O)O
Tekniske data
Kemisk navn eller materiale 3-(3,4,6-Trihydroxyphenyl)-alanine
InChI formel InChI=1S/C9H11NO5/c10-5(9(14)15)1-4-2-7(12)8(13)3-6(4)11/h2-3,5,11-13H,1,10H2,(H,14,15)/t5-/m0/s1
Mængde 250 mg
Formel vægt 213.0637