Få mere at vide
6-Hydroxy Bentazon-d7, TRC
Artikelnummer. 30405441
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30405441 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30405441 Leverandør Toronto Research Chemicals Leverandørnr. TRCH8065021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 6-Hydroxy Bentazon-d7
CAS 1330180-76-1
Molekylær formel C10H5D7N2O4S
Mængde 1 mg
Synonym 6-Hydroxy-3-(1-methylethyl-d7)-1H-2,1,3-benzothiadiazin-4(3H)-one 2,2-Dioxide, 6-Hydroxybentazone-d7
InChI formel InChI=1S/C10H12N2O4S/c1-6(2)12-10(14)8-5-7(13)3-4-9(8)11-17(12,15)16/h3-6,11,13H,1-2H3/i1D3,2D3,6D
SMIL [2H]C([2H])([2H])C([2H])(N1C(=O)c2cc(O)ccc2NS1(=O)=O)C([2H])([2H])[2H]
IUPAC navn 3-(1,1,1,2,3,3,3-heptadeuteriopropan-2-yl)-6-hydroxy-2,2-dioxo-1H-2λ^{6},1,3-benzothiadiazin-4-one
Molekylvægt (g/mol) 263.32
Formel vægt 263.0957