Få mere at vide
6-Hydroxy-3,4-dihydro-1H-quinolin-2-one, Mikromol™
Artikelnummer. 30052572
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30052572 100 mg
30051142 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30052572 Leverandør Mikromol Leverandørnr. MM0796.02
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
6-Hydroxy-3,4-dihydro-1H-quinolin-2-one
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 6-Hydroxy-3,4-dihydro-1H-quinoline-2-one
CAS 54197-66-9
Analyt- eller komponentnavne 6-Hydroxy-3,4-dihydro-1H-quinoline-2-one
Forsendelsestilstand Room Temperature
Molekylær formel C9 H9 N O2
Certificeringer/compliance ISO 17025
Synonym 6-Hydroxy-3,4-dihydro-2(1H)-quinolinone, 2(1H)-Quinolinone, 3,4-dihydro-6-hydroxy-Carbostyril, 3,4-dihydro-6-hydroxy- (7CI), Hydrocarbostyril, 6-hydroxy- (6CI), 2-Oxo-1,2,3,4-tetrahydroquinolin-6-ol, 3,4-Dihydro-6-hydroxy-2(1H)-quinolinone, 3,4-Dihydro-6-hydroxycarbostyril, 6-Hydroxy-1,2,3,4-tetrahydro-2-quinolinone, 6-Hydroxy-2-oxo-1,2,3,4-tetrahydroquinoline, 6-Hydroxy-3,4-dihydro-1H-quinolin-2-one, 6-Hydroxy-3,4-dihydro-2(1H)-quinolinone, 6-Hydroxy-3,4-dihydrocarbostyril, Cilostazol Related Compound A
InChI formel InChI=1S/C9H9NO2/c11-7-2-3-8-6(5-7)1-4-9(12)10-8/h2-3,5,11H,1,4H2,(H,10,12)
SMIL Oc1ccc2NC(=O)CCc2c1
IUPAC navn 6-hydroxy-3,4-dihydro-1H-quinolin-2-one
Molekylvægt (g/mol) 163.17
Mængde 100 mg
Formel vægt 163.0633 g/mol