Learn More
6*His, His-Tag Mouse, CoraLite 647, Clone: 1B7G5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL647-66005-100UL
Additional Details : Weight : 0.00020kg
Description
Protein tags are protein or peptide sequences located either on the C- or N- terminal of the target protein, which facilitates one or several of the following characteristics: solubility, detection, purification, localization and expression. His-tag is often used for affinity purification and binding assays. Expressed His-tagged proteins can be purified and detected easily because the string of histidine residues binds to several types of immobilized metal ions, including nickel, cobalt and copper, under specific buffer conditions. The His-tag antibody is a useful tool for monitoring of the His-tagged proteins, and recognizes His-tags placed at N-terminal, C-terminal, and internal regions of fusion proteins expressed in bacteria, insect, and mammalian cells.
Specifications
6*His, His-Tag | |
Monoclonal | |
2454 μg/mL | |
Mouse | |
Primary |
Immunofluorescence, Flow Cytometry | |
1B7G5 | |
CoraLite 647 | |
100 μL | |
Liquid |