Få mere at vide
6-Fluoro-3-(1-hydroxy-4-piperidyl)-1,2-benzoxazole, Mikromol™
Artikelnummer. 30052591
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30052591 25 mg
30051512 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30052591 Leverandør Mikromol Leverandørnr. MM0491.360025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
6-Fluoro-3-(1-hydroxy-4-piperidyl)-1,2-benzoxazole
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 6-Fluoro-3-(1-hydroxy-4-piperidyl)-1,2-benzoxazole
CAS 2985481-41-0
Analyt- eller komponentnavne 6-Fluoro-3-(1-hydroxy-4-piperidyl)-1,2-benzoxazole
Forsendelsestilstand Room Temperature
Molekylær formel C12 H13 F N2 O2
Certificeringer/compliance ISO 17025
Synonym 6-Fluoro-3-(1-hydroxy-4-piperidyl)-1,2-benzoxazole
InChI formel InChI=1S/C12H13FN2O2/c13-9-1-2-10-11(7-9)17-14-12(10)8-3-5-15(16)6-4-8/h1-2,7-8,16H,3-6H2
SMIL ON1CCC(CC1)c2noc3cc(F)ccc23
IUPAC navn 6-fluoro-3-(1-hydroxypiperidin-4-yl)-1,2-benzoxazole
Molekylvægt (g/mol) 236.24
Mængde 25 mg
Formel vægt 236.0961 g/mol