Få mere at vide
6-Ethylchenodeoxycholic Acid, TRC
Artikelnummer. 30133386
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133386 100 mg
30136601 50 mg
17930665 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30133386 Leverandør Toronto Research Chemicals Leverandørnr. TRCE899810100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 459789-99-2
Molekylær formel C26 H44 O4
Molekylvægt (g/mol) 420.63
Synonym Cholan-24-oic acid, 6-ethyl-3,7-dihydroxy-, (3α,5β,6α,7α)-, (3α,5β,6α,7α)-6-Ethyl-3,7-dihydroxycholan-24-oic acid, 3α,7β-Dihydroxy-6α-ethyl-5β-cholan-24-oic acid, 6-ECDCA, 6-Ethylchenodeoxycholic acid, 6α-Ethylchenodeoxycholic acid, INT 747, Obeticholic acid, Ocaliva
IUPAC navn (4R)-4-[(3R,5S,6R,7R,8S,9S,10S,13R,14S,17R)-6-ethyl-3,7-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid
SMIL CC[C@H]1[C@@H](O)[C@H]2[C@@H]3CC[C@H]([C@H](C)CCC(=O)O)[C@@]3(C)CC[C@@H]2[C@@]4(C)CC[C@@H](O)C[C@@H]14
Tekniske data
Kemisk navn eller materiale 6-Ethylchenodeoxycholic Acid
Mængde 100 mg
InChI formel InChI=1S/C26H44O4/c1-5-17-21-14-16(27)10-12-26(21,4)20-11-13-25(3)18(15(2)6-9-22(28)29)7-8-19(25)23(20)24(17)30/h15-21,23-24,27,30H,5-14H2,1-4H3,(H,28,29)/t15-,16-,17-,18-,19+,20+,21+,23+,24-,25-,26-/m1/s1
Formel vægt 420.324