Få mere at vide
6-Ethyl Guanine-d5, TRC
Artikelnummer. 30371791
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30371791 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30371791 Leverandør Toronto Research Chemicals Leverandørnr. TRCE9186525MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 6-Ethyl Guanine-d5
CAS 183558-84-1
Molekylær formel C7H4D5N5O
Mængde 5 mg
Synonym 6-Ethylguanine-d5, 6-Ethoxy-9H-purin-2-amine-d5, 2-Amino-6-ethoxypurine-d5, 2-Amino-6-ethoxypurine-d5, 6-Ethoxy-1H-purin-2-amine-d5, 6-O-Ethylguanine-d5, NSC 37368-d5, O6-Ethylguanine-d5
InChI formel InChI=1S/C7H9N5O/c1-2-13-6-4-5(10-3-9-4)11-7(8)12-6/h3H,2H2,1H3,(H3,8,9,10,11,12)/i1D3,2D2
SMIL NC1=NC(OC([2H])(C([2H])([2H])[2H])[2H])=C2NC=NC2=N1
IUPAC navn 6-(ethoxy-d5)-7H-purin-2-amine
Molekylvægt (g/mol) 184.21
Formel vægt 184.11