Få mere at vide
6-Despiperidinyl-6-(alogliptin-Namino-yl) Alogliptin, TRC
Artikelnummer. 30354761
Skift visning
Mængde:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30354761 250 mg
30363301 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30354761 Leverandør Toronto Research Chemicals Leverandørnr. TRCD297155250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1268836-55-0
Molekylær formel C31H30N8O4
Molekylvægt (g/mol) 578.62
Synonym 2-[[6-[(3R)-3-[[3-[(2-cyanophenyl)methyl]-1,2,3,6-tetrahydro-1-methyl-2,6-dioxo-4-pyrimidinyl]amino]-1-piperidinyl]-3,4-dihydro-3-methyl-2,4-dioxo-1(2H)-pyrimidinyl]methyl]-benzonitrile
IUPAC navn (R)-2-((6-(3-((3-(2-cyanobenzyl)-1-methyl-2,6-dioxo-1,2,3,6-tetrahydropyrimidin-4-yl)amino)piperidin-1-yl)-3-methyl-2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)methyl)benzonitrile
SMIL C(N1C(=CC(=O)N(C)C1=O)N2C[C@H](NC=3N(CC4=C(C#N)C=CC=C4)C(=O)N(C)C(=O)C3)CCC2)C5=C(C#N)C=CC=C5
Tekniske data
Kemisk navn eller materiale 6-Despiperidinyl-6-(alogliptin-Namino-yl) Alogliptin
Mængde 250 mg
InChI formel InChI=1S/C31H30N8O4/c1-35-28(40)14-26(38(30(35)42)18-23-10-5-3-8-21(23)16-32)34-25-12-7-13-37(20-25)27-15-29(41)36(2)31(43)39(27)19-24-11-6-4-9-22(24)17-33/h3-6,8-11,14-15,25,34H,7,12-13,18-20H2,1-2H3/t25-/m1/s1
Formel vægt 578.24