Få mere at vide
6-Desacetyl-6-bromo-N-Boc Palbociclib-d4, TRC
Artikelnummer. 30374724
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30374724 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30374724 Leverandør Toronto Research Chemicals Leverandørnr. TRCD44587410MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 6-Desacetyl-6-bromo-N-Boc Palbociclib-d4
CAS 2089333-16-2
Molekylær formel C9 H7 F3 O3
Mængde 10 mg
Synonym 4-[6-[(6-Bromo-8-cyclopentyl-7,8-dihydro-5-methyl-7-oxopyrido[2,3-d]pyrimidin-2-yl)amino]-3-pyridinyl]-1-piperazine-2,2,6,6-d4-carboxylic-d4 Acid 1,1-Dimethylethyl Ester, tert-Butyl 4-[6-(6-Bromo-8-cyclopentyl-5-methyl-7-oxo-7,8-dihydro-pyrido[2,3-d]pyrimidin-2-ylamino)-pyridin-3-yl]-piperazine-1-carboxylate-d4
InChI formel InChI=1S/C9H7F3O3/c10-9(11,12)6-2-1-3-7(4-6)15-5-8(13)14/h1-4H,5H2,(H,13,14)
SMIL OC(=O)COc1cccc(c1)C(F)(F)F
IUPAC navn 2-[3-(trifluoromethyl)phenoxy]acetic acid
Molekylvægt (g/mol) 220.145
Formel vægt 220.035