Få mere at vide
6-des-flouro-Fluticasone Furoate, TRC
Artikelnummer. 30358765
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30358765 10 mg
30357508 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30358765 Leverandør Toronto Research Chemicals Leverandørnr. TRCD39845010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 9-Fluoro-17Beta-(((fluoromethyl)sulfanyl)carbonyl)-11Beta-hydroxy-16Alpha-methyl-3-oxoandrosta-1,4-dien-17Alpha-yl furan-2-carboxylate
CAS 398454-97-2
Molekylær formel C27 H30 F2 O6 S
Mængde 10 mg
Synonym Androsta-1,4-diene-17-carbothioic acid, 9-fluoro-17-[(2-furanylcarbonyl)oxy]-11-hydroxy-16-methyl-3-oxo-, S-(fluoromethyl) ester, (11β,16α,17α)- (9CI), 9-Fluoro-17β-[[(fluoromethyl)sulfanyl]carbonyl]-11β-hydroxy-16α-methyl-3-oxoandrosta-1,4-dien-17α-yl fu
InChI formel InChI=1S/C27H30F2O6S/c1-15-11-19-18-7-6-16-12-17(30)8-9-24(16,2)26(18,29)21(31)13-25(19,3)27(15,23(33)36-14-28)35-22(32)20-5-4-10-34-20/h4-5,8-10,12,15,18-19,21,31H,6-7,11,13-14H2,1-3H3/t15-,18+,19+,21+,24+,25+,26+,27+/m1/s1
SMIL C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(OC(=O)c5occc5)C(=O)SCF
IUPAC navn [(8S,9R,10S,11S,13S,14S,16R,17R)-9-fluoro-17-(fluoromethylsulfanylcarbonyl)-11-hydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl] furan-2-carboxylate
Molekylvægt (g/mol) 520.585
Formel vægt 520.173