Få mere at vide
6-Demethyl Papaverine-d3, TRC
Artikelnummer. 30365204
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30365204 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30365204 Leverandør Toronto Research Chemicals Leverandørnr. TRCD23120210MG
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 6-Demethyl Papaverine-d3
CAS 1246815-25-7
Molekylær formel C19H16D3NO4
Mængde 10 mg
Synonym 1-[(3,4-Dimethoxyphenyl)methyl]-7-(methoxy-d3)-6-isoquinolinol, 7-(Methoxy-d3)-1-veratryl-6-isoquinolinol, 6-Desmethylpapaverine-d3, Isopacodine-d3, NSC 273754-d3
InChI formel InChI=1S/C19H19NO4/c1-22-17-5-4-12(9-19(17)24-3)8-15-14-11-18(23-2)16(21)10-13(14)6-7-20-15/h4-7,9-11,21H,8H2,1-3H3/i2D3
SMIL OC1=CC(C=CN=C2CC3=CC=C(OC)C(OC)=C3)=C2C=C1OC([2H])([2H])[2H]
Molekylvægt (g/mol) 328.38
Formel vægt 328.1502