missing translation for 'onlineSavingsMsg'
Learn More

6-Chloroimidazo[2,1-b]thiazole-5-sulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog Number p-7032284
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
150020-64-7
C5H2Cl2N2O2S2
257.103
MFCD00205139
APRATEMYDZDYJD-UHFFFAOYSA-N
6-chloroimidazo 2,1-b 1,3 thiazole-5-sulfonyl chloride, 6-chloroimidazo 2,1-b thiazole-5-sulfonyl chloride, 6-chloroimidazo 2,1-b thiazole-5-sulfonylchloride, 6-chloro-imidazo 2,1-b thiazole-5-sulfonyl chloride, imidazo 2,1-b thiazole-5-sulfonylchloride, 6-chloro, acmc-1c5af, 6-chloroimidazo 2,1-b 1,3 thiazole-5-sulfonylchloride, 6-chloro-5-imidazo 2,1-b thiazolesulfonyl chloride, 6-chloro-imidazo 2, 1-b thiazole-5-sulfonyl chloride, imidazo 2,1-b thiazole-5-sulfonylchloride,6-chloro
2799717
6-chloroimidazo[2,1-b][1,3]thiazole-5-sulfonyl chloride
C1=CSC2=NC(=C(N21)S(=O)(=O)Cl)Cl
This item is not returnable. View return policy
150020-64-7
C5H2Cl2N2O2S2
257.103
MFCD00205139
APRATEMYDZDYJD-UHFFFAOYSA-N
6-chloroimidazo 2,1-b 1,3 thiazole-5-sulfonyl chloride, 6-chloroimidazo 2,1-b thiazole-5-sulfonyl chloride, 6-chloroimidazo 2,1-b thiazole-5-sulfonylchloride, 6-chloro-imidazo 2,1-b thiazole-5-sulfonyl chloride, imidazo 2,1-b thiazole-5-sulfonylchloride, 6-chloro, acmc-1c5af, 6-chloroimidazo 2,1-b 1,3 thiazole-5-sulfonylchloride, 6-chloro-5-imidazo 2,1-b thiazolesulfonyl chloride, 6-chloro-imidazo 2, 1-b thiazole-5-sulfonyl chloride, imidazo 2,1-b thiazole-5-sulfonylchloride,6-chloro
2799717
6-chloroimidazo[2,1-b][1,3]thiazole-5-sulfonyl chloride
C1=CSC2=NC(=C(N21)S(=O)(=O)Cl)Cl

It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 138°C to 140°C
Quantity 1 g
UN Number UN3261
Sensitivity Moisture sensitive
Solubility Information Sparingly soluble in water.(0.26g/L) (25°C),
Formula Weight 257.12
Percent Purity 97%
Chemical Name or Material 6-Chloroimidazo[2,1-b]thiazole-5-sulfonyl chloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'