Få mere at vide
6-Chloro1,4-dihydro-1-methyl-4-phenyl-4-quinazolinol, TRC
Artikelnummer. 30360186
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30360186 10 mg
30354527 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30360186 Leverandør Toronto Research Chemicals Leverandørnr. TRCC37102510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 52395-03-6
Molekylær formel C15 H13 Cl N2 O
Molekylvægt (g/mol) 272.73
Synonym 6-Chloro-1,4-dihydro-4-hydroxy-1-methyl-4-phenylquinazoline, 6-chloro-1,4-dihydro-1-methyl-4-phenylquinazolin-4-ol
IUPAC navn 6-chloro-1-methyl-4-phenylquinazolin-4-ol
SMIL CN1C=NC(O)(c2ccccc2)c3cc(Cl)ccc13
Tekniske data
Kemisk navn eller materiale 6-Chloro-1,4-dihydro-4-hydroxy-1-methyl-4-phenylquinazoline
Mængde 10 mg
InChI formel InChI=1S/C15H13ClN2O/c1-18-10-17-15(19,11-5-3-2-4-6-11)13-9-12(16)7-8-14(13)18/h2-10,19H,1H3
Formel vægt 272.0716