Få mere at vide
6-Chloro1,4-dihydro-1-methyl-4-phenyl-4-quinazolinol, TRC
Artikelnummer. 30354527
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30354527 100 mg
30360186 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30354527 Leverandør Toronto Research Chemicals Leverandørnr. TRCC371025100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 52395-03-6
Molekylær formel C15 H13 Cl N2 O
Molekylvægt (g/mol) 272.73
Synonym 6-Chloro-1,4-dihydro-4-hydroxy-1-methyl-4-phenylquinazoline, 6-chloro-1,4-dihydro-1-methyl-4-phenylquinazolin-4-ol
IUPAC navn 6-chloro-1-methyl-4-phenylquinazolin-4-ol
SMIL CN1C=NC(O)(c2ccccc2)c3cc(Cl)ccc13
Tekniske data
Kemisk navn eller materiale 6-Chloro-1,4-dihydro-4-hydroxy-1-methyl-4-phenylquinazoline
Mængde 100 mg
InChI formel InChI=1S/C15H13ClN2O/c1-18-10-17-15(19,11-5-3-2-4-6-11)13-9-12(16)7-8-14(13)18/h2-10,19H,1H3
Formel vægt 272.0716